Spécifications
- Packaging:Glass Bottle
- Description:M-GLAS™ Liquid Cover Glass
- Size:500 mL
- Cat. no.:CA1.03973.0001
- Supplier No.:1.03973.0001
Spécifications
A propos de cet article
M-GLAS® should be used instead of a cover glass for the homogeneous mounting of cytological specimens
Place a few drops of the mounting medium on the specimen and spread evenly. When the solvent has evaporated, a firm protective surface coating will evolve. (Refractive index (20°C): 1.490 - 1.500). It is registered as IVD and CE product.