A propos de cet article
Dispenser and Pump Syringes from SGE incorporate Diamond Technology, taking advantage of new and innovative technologies and bringing those benefits to your laboratory. For over 50 years, SGE Analytical Science has been designing and manufacturing syringes for a variety of instruments and customer applications. At the heart of your pump or dispenser, and combined with the rest of the system components, syringes must provide the superior performance you demand and rely upon. Designed and optimized with outstanding quality and precise fit, SGE syringes provide greater reliability for applications run on your systems.
Dispenser and Pump Syringes from SGE incorporate Diamond Technology, taking advantage of new and innovative technologies and bringing those benefits to your laboratory. For over 50 years, SGE Analytical Science has been designing and manufacturing syringes for a variety of instruments and customer applications. At the heart of your pump or dispenser, and combined with the rest of the system components, syringes must provide the superior performance you demand and rely upon. Designed and optimized with outstanding quality and precise fit, SGE syringes provide greater reliability for applications run on your systems.
- Improved accuracy and precision
- Robust and durable for longer life, less maintenance downtime and reduced maintenance costs
- Do not contaminate the aspirated fluid
- Designed to perfectly fit your instrument, making them easy to use and install
Dispenser systems. 60mm stroke. Luer Lock or 1/4-28 UNF thread connection. 6-32 UNC thread in plunger button (CX has a button stop).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 14233-546
- 14225-328
- 14225-358
- 14225-274
- 14225-410
- 14225-420
- 14225-390
- 14225-376
- 14225-350
- 14225-292
- 14225-402
- 75815-604
- Operating temperature range:
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C
- 10 to 40°C