Accessories for Handheld Fluorimeter, ANDalyze
Fournisseur: ANDalyze Inc
Certificats
Certificats
A propos de cet article
Customers Who Bought This Also Bought
Sodium hydroxide 0.1 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$194.14 to $694.12
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Fournisseur: MilliporeSigma
$1,313.27 to $5,149.78
Sulfuric acid 1.0 N in aqueous solution, VWR Chemicals BDH®
Fournisseur: VWR International
$170.25 to $676.42
TruBond™ Adhesion Slides, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
$123.32 to $123.32