To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Branched Chain Keto Acid Dehydrogenase E1, Beta Polypeptide
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:BCKDHB
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- Size:150 µL
- Western blot:Yes
- Storage buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage temperature:Store at –20 °C
- Immunogen:BCKDHB
- Purification:Antigen affinity purification
- Cat. no.:10156-008
- Supplier No.:13685-1-AP
Spécifications
A propos de cet article
The BCKDHB antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human BCKDHB. This antibody recognizes human,mouse,rat antigen. The BCKDHB antibody has been validated for the following applications: ELISA, WB, IHC analysis.
WB 1 : 1000
Type: Primary
Antigen: BCKDHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat