ContactUsLinkComponent
Potato Dextrose Agar, with 3% Agar, HiMedia Laboratories
Potato Dextrose Agar, with 3% Agar, HiMedia Laboratories
# de catalogue CA61000-212
Fournisseur:  HiMedia
Numéro CAS:  
Potato Dextrose Agar, with 3% Agar, HiMedia Laboratories
# de catalogue CA61000-212
Fournisseur:  HiMedia
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • pH:
  • Description:
    Potato Dextrose Agar with 3% Agar
  • Cat. no.:
    CA61000-212
  • Supplier No.:
    M937-500G
Spécifications
A propos de cet article
For isolation and cultivation of fungi-yeasts and moulds from dairy and food products.
  • Isolate and cultivate fungi yeasts and moulds
  • Recommended by the APHA and FDA
  • Agar enhances sporulation in the medium
Potato Dextrose Agar is recommended by APHA and FDA for plate counts of yeasts and moulds in the examination of foods and dairy products. Incorporation of 3% agar enhances sporulation in the medium
Customers Who Bought This Also Bought