Spécifications
- pH:—
- Description:Potato Dextrose Agar with 3% Agar
- Cat. no.:CA61000-212
- Supplier No.:M937-500G
Spécifications
A propos de cet article
For isolation and cultivation of fungi-yeasts and moulds from dairy and food products.
- Isolate and cultivate fungi yeasts and moulds
- Recommended by the APHA and FDA
- Agar enhances sporulation in the medium
Potato Dextrose Agar is recommended by APHA and FDA for plate counts of yeasts and moulds in the examination of foods and dairy products. Incorporation of 3% agar enhances sporulation in the medium