GC Ferrules, Graphite, Trajan Scientific and Medical
Fournisseur: Trajan Scientific and Medical
Certificats
A propos de cet article
Your chromatography analysis does not end with the selection of the GC column. The combination of components selected for your instrument also make an important contribution to successful separations. SGE brings technology and chromatography expertise to improve your chromatography. SGE manufactures an array of connection types for GC use. Poorly defined or selected connections can lead to an increase in dead volume, leaks from cycled temperatures and mismatched tubing sizes.
- Easy to use
- Forms a reliable seal
- Reusable
- Soft material
- Forms a soft grip to the capillary column
- Low emissions
Column to injector and column to FID connections. Porous to oxygen, unsuitable for connecting columns to mass spectrometers.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: A variety of hole sizes are available for 1/16, 1/8 and 1/4" fittings. 450 ºC temperature limit. A variety of profiles are available, including short, blank, 2 and 3-hole ferrules.
Customers Who Bought This Also Bought