A propos de cet article
Features and Benefits–SGE’s unique energized plunger tip design ensures a gas tight seal is consistently maintained providing outstanding performance (%RSD) with temperature cycling.
Features and Benefits–SGE’s unique energized plunger tip design ensures a gas tight seal is consistently maintained providing outstanding performance (%RSD) with temperature cycling.
- Outstanding reproducibility maintained during temperature cycling
- Highest operating temperature
- Save time and improve analysis workflow
The PTFE plunger tip in Diamond Headspace Syringes enables the use of a broader range of samples and solvents compared with polymers used in plungers of other high temperature syringes, this provides greater flexibility for your headspace analyses and a longer lasting syringe.
No dead space. GC Headspace injections.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 89236-288
- 89236-286
- 89236-278
- 89236-276
- Accuracy:
- ± 1 % (displaced volume), accuracy and reproducibility
- ± 1 % (displaced volume), accuracy and reproducibility
- ± 1 % (displaced volume), accuracy and reproducibility
- ± 1 % (displaced volume), accuracy and reproducibility
- Needle length:
- 5.6 cm
- 5.6 cm
- 5.6 cm
- 5.6 cm
- Point style:
- Side-hole
- Side-hole
- Side-hole
- Side-hole
- Temperature range:
- 25-150°C
- 25-150°C
- 25-150°C
- 25-150°C