Graduated Midget Bubblers, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Micro impinger designed for sampling of small volumes of air at low jet velocity, as per EPA Method 6
Impinger bottle is graduated from 5 to 30 ml in 5 ml subdivisions. Both the bottle and stopper have 24/40 standard taper joints. The impinge tip has an extra coarse fritted gas dispersion tube which enhances absorption by trapping the liquid.
Customers Who Bought This Also Bought