A propos de cet article
The TouchNTuff® 73-500 neoprene gloves deliver superior sensitivity and comfort in sterile, aseptic environments.
- These sensitive neoprene gloves are a new feel in double-donning and dry applications
- TouchNTuff® single-use gloves provide superior tactility
- High resilience in wet and dry industrial, lab, and controlled environments
Gloves are packed in a controlled environment, in a cleanroom-compatible packaging material, which is printed with IPA-resistant ink.
Customers need to make their own determination based on cleanliness standards. Ansell expects this glove should be suitable for ISO Class 5 or higher cleanroom environments.
The exceptionally thin polychloroprene formulation reduces the risk of Type I or IV allergies to the wearer caused by natural rubber latex or other synthetic gloves. The sensitive neoprene gloves offer users splash resistance against a broad range of chemicals. SureFit Technology™ eliminates 'cuff roll down' by maintaining gloves securely around the arm, ensuring the efficacy of the gloves as a barrier protection. Gloves are ergonomically-designed and hand specific to enhance fit and comfort, and to reduce hand fatigue and stress.
Spécifications
- Catalog No:
- 10055-027
- 10055-025
- 10055-023
- 10054-587
- 10054-585
- 10054-595
- 10054-593
- 10054-589
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Rolled, Beaded
- Rolled, Beaded
- Rolled, Beaded
- Rolled, Beaded
- Rolled, Beaded
- Rolled, Beaded
- Rolled, Beaded
- Rolled, Beaded