A propos de cet article
SGE has been making precision chromatography syringes for over 50 years
Features and Benefits–Unique plunger design minimizes bending and seizing.
Features and Benefits–Unique plunger design minimizes bending and seizing.
Metal plungers are individually fitted to the syringe glass barrel for a perfect “feel”, optimized life with minimal carry over, a liquid tight seal between the barrel and plunger and excellent performance.
PTFE tipped replaceable plunger syringes reduce the risk of plunger seizing when working with dirty samples, reducing instrument downtime, and minimizing the risk of instrument shutdown during the processing of large batches of samples.
Accuracy and Reproducibility is ± 1 % (displaced volume). Temperature Range is 5-70 °C Fixed Needle, 5-120 °C Removable Needle.
Our range of HPLC autosampler syringes are designed and tested to meet critical autosampler specifications.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 89238-048
- 60362-198
- 14225-316
- 14233-484
- 89238-046
- 89238-040
- 60362-204
- 89238-036
- 89238-052
- Gauge:
- 22 gauge
- 22 gauge
- 22 gauge
- 22 gauge
- 22 gauge
- 22 gauge
- 22 gauge
- 22 gauge
- 22 gauge
- Needle length:
- 5.1 cm
- 5.1 cm
- 5.1 cm
- 5.1 cm
- 5.1 cm
- 5.1 cm
- 5.1 cm
- 5.1 cm
- 5.1 cm
- Tip:
- LC
- LC
- LC
- LC
- LC
- LC
- LC
- LC
- LC