HyFlex® 74-045 Lightweight Dyneema® Gloves, White, Ansell
Catalog # 80092-796
Supplier: Ansell Healthcare
Specifications
- Glove Color:White
- Glove size:XS
- Ambidextrous/Hand-specific:Hand-specific
- Length:21.6 cm (8 1/2")
- Liner material:Dyneema®
- ASTM No.:254X
- Cat. no.:80092-796
- Supplier No.:222161
Specifications
About this item
See how a lightweight glove can deliver high-strength protection with these gloves that are specifically designed for the food processing industry.
- Exceptional cut protection
- Can be worn under other gloves
- Lightweight
The Dyneema® core construction with high-strength outer-wrap fibers offers excellent protection from cuts.
Use in the fishing, food processing, and food service industries. Uses include beef, pork, poultry, and fish processing; cleaning and changing stationary slicer blades in delis and supermarkets; as a liner under work gloves for applications where extra protection is required; kill and processing areas; and knife hand applications in meat processing.
May be used as a liner under another glove. Tuff-Cuff™ adds extra wrist protection and comfort.
Certifications: ANSI level 4 cut protection. ANSI level 3 abrasion protection.
Packaging: Gloves are packaged 12 each per case.
Customers Who Bought This Also Bought