A propos de cet article
ESD barrier lab coats are made with a fluid-resistant material consisting of 99% polyester and a 1% carbon stripe for electrostatic dissipation (ESD). These barrier coats are ideal for use in medical, healthcare, veterinary, and food processing applications.
- One breast pocket
- Two oversized waist pockets
- Cuffed sleeves
- Six snap closure
- Side seam openings for easy access to pants pockets
These coats are extremely durable and will hold up to repeated industrial launderings.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Catalog No:
- 76422-842
- 76422-844
- 10215-534
- 76422-846
- 10215-536
- 76422-826
- 10215-538
- 76422-834
- 10215-540
- 10215-542
- 76422-836
- 76422-838
- 10215-525
- 76422-840
- 10215-532
- 76422-828
- 76422-830
- 10215-527
- 10215-531
- 10215-529
- 76422-832
- Garment application:
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Medical, Food, Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- 99% polyester, 1% carbon stripe
- Weight:
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.
- 3.4 oz./sq. yd.