AccuTarget™ Control siRNA Set, Bioneer
Fournisseur: Bioneer
Certificats
A propos de cet article
AccuTarget™ Control siRNAs are a wide range of positive control siRNAs, negative control siRNAs and control siRNA sets
Control siRNA can be fluorescently labeled for monitoring transfection efficiency.
Customers Who Bought This Also Bought