A propos de cet article
AccuTarget Predesigned siRNA Library offers 32,000 predesigned siRNAs for more than 44,000 target genes from Human, Mouse and Rat.
AccuTarget Predesigned siRNA Library offers 32,000 predesigned siRNAs for more than 44,000 target genes from Human, Mouse and Rat. The gene of interest in siRNA library could be search by Gene ID, Symbol, Synonyms, Description, or Accession Number. Then, the predicted siRNA knockdown efficiency could be calculated by choosing the guaranteed yield and purification level. The qPCR primers also can be ordered for siRNA knockdown validation.
- High siRNA knockdown rates
- 2 of three siRNAs suppress target mRNA levels > 80%
- Unique design algorithm
- aximizes siRNA knockdown while minimizing off target effects Competitive pricing: Great value for your research dollar
- siRNA is supplied as a duplex
- siRNA Knockdown efficiency data
- NC (AccuTarget Negative Control) and siRNA concentration at 100 nM
- Transfection efficiency data
- PC (AccuTarget GAPDH/ GFP/Luciferase siRNA) and NC (AccuTarget Fluoresceinlabeled Negative Control)
The gene of interest in siRNA library could be search by Gene ID, Symbol, Synonyms, Description, or Accession Number. Then, the predicted siRNA knockdown efficiency could be calculated by choosing the guaranteed yield and purification level. The qPCR primers also can be ordered for siRNA knockdown validation.
AccuTarget validated siRNA are 21 pre-validated libraries most with knockdown rates >80%. Validated siRNAs can be ordered at 10, 20, 50, or 100 nmole scale.
AccuTarget siRNA library can be used in a variety of RNAi experiments like functional genomics and proteomics research, gene expression studies and array analysis.