SilFlow™ Gas Chromatography Fittings, Trajan Scientific and Medical
Fournisseur: Trajan Scientific and Medical
Certificats
A propos de cet article
SGE understands today’s chromatographers need to move from tubing based flow systems to planar microchannel systems to deliver flexible chromatography solutions
- Chemically inert
- Low dead volume
- Superb operational stability
- Easy to install
- Leak free
These various fittings can be used for backflushing, column splitting, detector splitting, heart cutting, deans' switching and column switching. They are for use in multidimensional GC and anywhere a tee-piece has traditionally been used.
SilFlow™ is an innovation in design and fabrication resulting in a highly efficient and reliable micro fluidic platform that improves your GC connectivity to enable maximum chromatography performance.
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: Various configurations are available with ports to suit 0.25/0.32 mm and 0.53 mm I.D. columns, fused silica and 1.1 mm O.D. stainless steel tubing.
Customers Who Bought This Also Bought