A propos de cet article
Trypsin Delarco assists in the process of cell culture and transfection.
- With HBSS without calcium and magnesium
- Without Phenol Red
Trypsin is a proteolytic enzyme used to detach adherent cells from culture vessel surfaces. Trypsin delarco is used in cell passaging, to ensure stable products.
Sterile Filtered
Concentration: 0.1 mM disodium EDTA
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 36 months