A propos de cet article
L-glutamine is a cell culture supplement, and an essential amnio acid. L-glutamine is required in cell culture for cellular growth.
- 100X concentration
L-glutamine is used in cell culture as an energy source in addition to carbohydrates (i.e. glucose, sucrose, maltose, etc.). L-glutamine participates in the formation of purine and pyrimidine nucleotides, amino sugars, gluthione, L-glutamate and other amino acids, as well as in protein synthesis and glucose production.
Sterile Filtered
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: 240 months