A propos de cet article
MEM NEAA contains the same non-essential amino acids found in the standard MEM, at a strength of 100. MEM NEAA 100X can be used to enhance cell growth and prolong the viability of cells in culture. MEM NEAA 100X is commonly used as a supplement to minimal basal medium, and does not dilute the components already in the cell culture medium.
- 100X of Non-essential amino acids
MEM NEAA 100X is useful for supplementing cell culture medium to maintain cell culture growth and study cellular protein metabolism, and optimizes performance in the in vitro hematopoietic differentiation of mouse embryonic stem cells.
Sterile Filtered
Harmonization Code: 3822.00.5090
ECCN: EAR99