Tryptic Soy Agar Plates with 5% Sheep Blood, Quality Biological
Catalogue # 10128-598
Fournisseur: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Numéro CAS:
Spécifications
- Description:TSA Plates with 5% Sheep Blood
- Size:25 mL
- Storage conditions:2°C to 8°C
- Sterility:Autoclaved
- Shipping conditions:Ambient Temperature
- Cat. no.:10128-598
- Shelf Life:6 Months
- Supplier No.:441-120-231
Spécifications
À propos de cet article
Trypticase Soy Agar with 5% Sheep Blood is used for cultivating fastidious microorganisms and for the visualization of hemolytic reactions produced by many bacterial species
Grow a wide variety of fastidious organisms on TSA with Sheep Blood, a general purpose non-selective medium with the addition of sheep blood in the formulation.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only