Ampicillin 100 mg/mL ready-to-use, sterile filtered
Catalog # 10128-594
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number: 69-53-4
Specifications
- Size:1 ml
- Cat. no.:10128-594
- Supplier No.:351-344-731
Specifications
Specification Test Results
- Ampicillin 100 mg/mL ready-to-use, sterile filtered:
Specification Test Results
About this item
:Ampicilline
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Pt ébul.:|Point d'ébullition:
:Pt Fusion:|Point de fusion:
:Pt Eclair:|Point eclaire:
:M.W. 349,4 g/mol
:Ampicillin
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 349.4 g/mol
Storage Temperature:Freezer
:69-53-4
:C₁₆H₁₉N₃O₄S
:MFCD00005175
Customers Who Bought This Also Bought