About this item
Bisulfateless Brucella Plate (BLBB) is applicable for use in the primary isolation, quantitation, and partial identification of obligately anaerobic microorganisms from clinical specimens.
- Special formulation without bisulfate
BLBB with GSS Supplements is a non-selective medium suitable for the growth of aerobic and microaerophilic bacteria when incubated under the appropriate conditions. BLBB Plates are useful for the enrichment, isolation and cultivation of Brucella species.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: NA