A propos de cet article
LB Agar Plates are used for the growth and maintenance of E. coli strains used in molecular microbiology procedures.
The following variations of LB Agar Plates are nutritionally rich media designed for growth of pure cultures of recombinant strains. LB Agar plates are suitable for propagation of E. Coli and other bacteriological stocks.
UNSPSC: 41106200
Harmonization Code: 3822.00.5090
ECCN: EAR99
Max Shelf Life: NA