Pre-washed Pre-Weighed Glass Microfiber Filter, Paper Grade 169, Ahlstrom
# de catalogue 10199-602
Fournisseur: Ahlstrom
Spécifications
- Flow rate:Fast
- Size:4.7 cm dia.
- Retention:1.0 microns
- Load capacity:Very High
- Cat. no.:10199-602
- Supplier No.:169W-0470
Spécifications
A propos de cet article
Grade 169 is a microfiber glass grade formulated for product stability when heated over 500 °C as required for testing for volatile solids in STM 2540.
In this item each filter is washed and weighed and comes in an aluminum pan with the weight printed on the side for ease of use.
Customers Who Bought This Also Bought