Glass Microfiber Filter Paper, Grade 169, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Grade 169 is a microfiber glass grade formulated for product stability when heated over 500 degrees Centrigrade, as required for testing for volatile solids in STM 2540.
Spécifications
- Catalog No:
- 10199-608
- 10199-610
- 10199-612
- 10199-614
- 10199-616
- 10199-604
- 10199-626
- 10199-606
- 10199-628
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Retention:
- 1.0 microns
- 1.0 microns
- 1.0 microns
- 1.0 microns
- 1.0 microns
- 1.0 microns
- 1.0 microns
- 1.0 microns
- 1.0 microns