Glass Microfiber Filter Paper, Grade 121, Ahlstrom
Fournisseur: Ahlstrom
Certificats
A propos de cet article
Grade 121 is a very thick binder-free microfiber glass filter paper that offers finer particle retention than Ahlstrom 111 with higher wet strength and loading capacity.
Spécifications
- Catalog No:
- 47745-794
- 47749-538
- 47749-540
- 47745-790
- 47749-534
- 47745-792
- 47749-536
- 47749-532
- 47749-542
- 38010-436
- 38010-429
- 38010-418
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Retention:
- 1.2 microns
- 1.2 microns
- 1.2 microns
- 1.2 microns
- 1.2 microns
- 1.2 microns
- 1.2 microns
- 1.2 microns
- 1.2 microns
- 1.2 microns
- 1.2 microns
- 1.2 microns