A propos de cet article
The patented RXPert Single Filtered Balance System is a Class I Enclosure that meets the USP<800> requirements for non-sterile compounding
The RXPert features a space-saving design constructed of a static-dissipative powder-coated aluminum frame, steel rear plenum and baffle, and tempered safety glass. This balance enclosure includes an all-in-one factory-installed canopy, and damper allows for thimble ducting to the outside. The true bag-in/bag-out HEPA filter disposal system allows for safe filter replacement.
The HEPA filter is easily accessed to allow the entire surface to be scanned for leak detection. It includes a built-in blower with dynamically-balanced, vibration-isolated motorized impeller and speed control. The blower has a 40% reserve so that the speed may be adjusted as needed by a certified technician as the filter loads.
Optional filtration kits and thimble ducting kits are available to convert existing XPert Balance Filtered Systems, XPert Bulk Powder Filtered Systems, and RXPert Single Filtered Balance Systems. Double Filtered models requiring no ductwork are also available.
It provides user protection by keeping powders and particulates contained during hazardous drug manipulation. It features durable glass and steel construction, ergonomic air foil with aerodynamic Clean-Sweep airflow openings, and deep interior of 23.4" or 30.0" depending on the model.
Spécifications
- Catalog No:
- 10216-844
- 10216-846
- 10216-848
- 10216-850
- 10216-852
- 10830-816
- 10830-814
- 10830-812
- 10830-810
- 10830-818
- 10830-800
- 10830-808
- 10830-806
- 10830-804
- 10830-802
- 10216-854
- 10216-856
- 10216-858
- 10216-860
- 10830-792
- 10830-790
- 10830-788
- 10830-822
- 10830-820
- 10830-786
- 10830-826
- 10830-824
- Environmentally Preferable:
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Sustainable Packaging, Sustainable Materials
- Electrical:
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz
- 115V, 60Hz