Spécifications
- Description:Optical Self Stirring BOD Probe with Temperature Sensor, Guarded Cap
- Temperature accuracy:+/-0.2ºC
- Temperature resolution:0.1ºC
- Temperature range:Ambient 10 to 40ºC (50 to 104ºF)
- DO resolution:0.01 mg/L; 0.1%
- DO concentration:0 to 50 mgL
- DO saturation:0 to 500%
- DO accuracy:0 to 20 mg/L, +/-0.1 mg/L or +/-1% of reading, whichever greater; 20 to 50 mg/L, +/-10% reading
- Response time:95% in 22 seconds with stirring; 95% in 40 seconds without stirring
- Cat. no.:10066-706
- Cable Length:2 m (6.5')
- Supplier No.:626500
Spécifications
A propos de cet article
The IDS ProOBOD® self-stirring optical BOD probe for use with the YSI MultiLab. Designed to fit into standard 300 mL BOD bottles.
- Smart sensor retains calibration in the probe; IDS = Intelligent Digital Sensor
- User-replaceable, low-maintenance sensor cap with 1-year life
- Guarded sensor - the sensor guard provides protection of the sensor tip when taking the BOD probe in and out of BOD bottles
- Probe body tapered to fit into a standard 300 mL BOD bottle
- Extremely quiet operation
- No warm-up time required
- Self-stirring to provide representative sample and increase response times
IDS sensors automatically store their unique serial number and calibration data. In addition, they also digitally process the measurement signal. The IDS sensors can be moved from instrument to instrument and maintain their calibration data and transmit that information to the new instrument.
Designed for use with all models of the YSI MultiLab, including the 4010-1W, 4010-2W, and 4010-3W.
Temperature compensation: Ambient 10 to 40ºC (50 to 104ºF)