ChemiWriter ECL, Azure Biosystems
Catalog # 10304-282
Supplier: Azure Biosystems
Product available on GSA Advantage®
Specifications
- Description:ChemiWriter ECL
- Cat. no.:10304-282
- Supplier No.:AC2146
Specifications
About this item
Write on your blot with the "ink" in the ChemiWriter ECL, and once you add HRP substrate, your writing will glow like chemiluminescence.
Draw on your visible marker, and you will not need to take a marker image to see your molecular weight ladder on your blots.
Customers Who Bought This Also Bought