A propos de cet article
Delivering bare-handed dexterity with the protection of a glove, these products provide a relaxed, ergonomic fit.
- Durable construction
- Machine washable
- Maximum dexterity and sensitivity
- Relaxed, ergonomic fit
Gloves have performance characteristics suited to a wide variety of applications and work environments.
The glove’s solid gray polyurethane coating hides soil, providing longer wear and reducing overall replacement costs. The durable gloves can be machine washed and reused with minimal shrinking and excellent recovery. Maximum dexterity and sensitivity for jobs that require handling or assembling small parts.
Diverse HyFlex® styles provide the best-in-class comfort, protection and dexterity required for the precise manipulation of small parts, controlled operation of power tools and gear, and direct handling of heavy parts and materials.
Use in the automotive and transportation, machinery and equipment, and maintenance industries. Uses include assembly of small pieces such as small bearings, joints and springs.
Spécifications
- Catalog No:
- 80092-480
- 80092-478
- 80092-472
- 80092-476
- 80092-474
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Finish:
- Palm Coated
- Palm Coated
- Palm Coated
- Palm Coated
- Palm Coated
- Cuff style:
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- Knit Wrist
- EN norm:
- 3131
- 3131
- 3131
- 3131
- 3131