BP5MS GC Capillary Column, SGE
Fournisseur: Trajan Scientific and Medical
Certificats
A propos de cet article
Innovative phase technologies and fused silica production capabilities provide end-to-end separation solutions for applications such as ultra trace analyses, pesticides and herbicides, hydrocarbons, solvents, phenols, amines, GC/MS and other specific detector applications.
- For your 5% MS analysis
- Optimized silphenylene for general purpose MS analyses
- Low bleed
- Great inertness
- High temperature
- Non-polar
Operating temperature from –40 to 330/350 °C.
Certifications: RoHS compliant per Directive 2011/65/EU
Ordering information: These 5% Phenyl Polysilphenylene-siloxane columns are suitable replacements for DB-5ms, ZB-5ms, Rxi-5Sil, MS, VF-5ms, CP-Sil 8 CB.
Spécifications
- Catalog No:
- 89232-102
- 89232-104
- 10799-664
- Description:
- GC Capillary Column, BP5MS
- GC Capillary Column, BP5MS
- GC Capillary Column, BP5MS
- Temperature limit:
- -40 to 330/350°C
- -40 to 330/350°C
- -40 to 330/350°C