Extra Bracket for BradyGlo™ Sign, Brady
# de catalogue CA89007-832
Fournisseur: Brady Worldwide
Spécifications
- Color:White
- Description:Extra Bracket for BradyGlo™ Sign, Glow-in-the-Dark BradyGlo™ Photoluminescent
- Material:Steel
- Cat. no.:CA89007-832
- Supplier No.:80756
Spécifications
A propos de cet article
Extra Bracket for BradyGlo™ Sign.
- Abrasion and chemical resistant
- Surface mounted
Customers Who Bought This Also Bought