In-Line Refractometer, PRM-100[alpha], ATAGO®
# de catalogue 89187-070
Fournisseur: Atago
Spécifications
- Measurement range (°Brix):1.32000–1.55700 nD 0.00–100.00% Brix
- Measurement range:1.32000–1.55700 nD 0.00–100.00% Brix
- Resolution:0.10% or 0.01% Brix (By selection)
- Model:PRM-100[alpha]
- Cat. no.:89187-070
- L×W×H:19.2 x 10 x 24 cm (8 x 39/10 x 91/2")
- Accuracy:±0.00010 nD ±0.005% brix
- Weight:3.3 kg (7.27 lbs.)
- Supplier No.:3574
Spécifications
A propos de cet article
In-line Refractometers designed for the measurement of refractive index, concentration and Brix scale.
- Wider Brix range
- Easy to read color display panel
This model consists RS-232C output for connection to a computer.
Ordering information: Supplied with a one-year manufacturer’s warranty. A calibration certificate is available for each unit. Contact your Avantor representative for information.
Customers Who Bought This Also Bought