Methyl Deactivated Fused Silica Capillary Tubing, SGE
Fournisseur: Trajan Scientific and Medical
Certificats
A propos de cet article
SGE's Methyl Deactivated Fused Silica Capillary Tubing is available in a range of sizes.
The tubing is chemically inert and thermally stable up to 380 °C (716 °F). It is ideally suited for use with capillary GC guard columns and transfer lines.
Certifications: RoHS compliant per Directive 2011/65/EU.
Customers Who Bought This Also Bought