Extra-Large Enclosed Lockout Station (Station Only), Brady Worldwide®
# de catalogue CA89007-240
Fournisseur: Brady Worldwide
Spécifications
- Color:Red on White
- Description:Extra-Large Enclosed Lockout Station (Station Only)
- Material:Polystyrene
- Size:30"H x 25"W x 5.25"D
- Cat. no.:CA89007-240
- Supplier No.:50995
Spécifications
A propos de cet article
Constructed of sturdy white polystyrene with predrilled holes for easy wall mounting
Pegboard-style holes for hooks are drilled into the back (8 hooks supplied standard). Built-in lock rail holds 20 to 25 locks, depending on lock size. Components sold seperately.
Customers Who Bought This Also Bought