Vacuum Pump System with Wireless Remote, Model SC920, KNF Neuberger
# de catalogue 97014-790
Fournisseur: KNF Neuberger Inc.
Spécifications
- Protection class:IP20 enclosure rating
- Flow rate:20 L/min.
- Model:SC920G
- Electrical:115 V, 60 Hz
- Final vacuum:1.5 torr
- Dimensions:366×294×423 mm
- Cat. no.:97014-790
- Supplier No.:USC920
Spécifications
A propos de cet article
Delivers precise vacuum with easy, safe control and monitoring from anywhere in the lab.
- Intuitive interface with touch controls and rotation knob
- Menus include unit of measurement, language, mode, parameters
- Graphic display shows process variables
- Evacuation (regulated pumping)
- Controlled emptying of a chamber
- Pressure Control; system controls pressure to set value (constant pressure)
- Automatic; system automatically finds vapor pressure of sample and adjusts process pressure accordingly
- Individual Pressure Function; system controls process according to user-defined pressure curve
- REPEAT function allows successive repetitions of programmed curve
- Manual process control can be selected at any time during an active process
- PC control Windows-based software (included) enables control via external PC
- Software supports display of pressure curves as a chart
- Setpoints can be saved
- Data can be exported to spreadsheets or text files
- USB interface
Oil-free vacuum system supplies clean, reliable vacuum source without hassles of handling pump oils or tainted aspiration water. Four operating modes make this a fast, versatile device for a wide range of pumping applications. Intelligent control system monitors actual pressure; system compensates for fluctuation. Low-speed start. Patented diaphragm stabilization system ensures high speeds at low pressures for faster processing times. System stops when setpoint pressure is achieved; cycles ON/OFF intermittently to stabilize pressure. Chemical-resistant gas carrying parts. Quiet operation.
Customers Who Bought This Also Bought