Lab-Sorb™ for Liquid Disposal, Diversified Biotech
Supplier: Diversified Biotech
Certificates
About this item
- Converts liquid waste to solid waste in seconds
- Lab-Sorb granules can rapidly absorb aqueous or mixed aqueous and organic solvent waste
- One 16 oz. jar can absorb up to 50 liters of liquid
Frequently Bought Together