Spécifications
- Description:Handheld Fluorimeter, Black
- Cat. no.:10204-980
- Supplier No.:AND002
Spécifications
A propos de cet article
The AND1100 Fluorimeter tests for metals in water in two steps and less than two minutes, significantly reducing the test expenditures, complexity and wait times experienced in traditional tests. Powered by DNA, it is a breakthrough in streamlining the water analysis and reporting processes. The AND1100 Fluorimeter is a platform centric portable device for heavy metals testing for drinking water and industrial water supplies. It uses an extremely sensitive measurement technique in tandem with ANDalyze sensor kits to determine the toxicity levels of heavy metal contamination. Sensor kits are available for lead, uranium, copper, mercury, zinc, and cadmium; with more to come.
The AND1100 Fluorimeter tests for metals in water in two steps and less than two minutes, significantly reducing the test expenditures, complexity and wait times experienced in traditional tests. Powered by DNA, it is a breakthrough in streamlining the water analysis and reporting processes. The AND1100 Fluorimeter is a platform centric portable device for heavy metals testing for drinking water and industrial water supplies. It uses an extremely sensitive measurement technique in tandem with ANDalyze sensor kits to determine the toxicity levels of heavy metal contamination. Sensor kits are available for lead, uranium, copper, mercury, zinc, and cadmium; with more to come.
- One-button push for metals detection and analysis
- Usable heavy metals data available quickly
- Pairs with ANDalyze sensor kits, available for a variety of metals parameters
- No Special knowledge of chemical testing required
- Data Capture and Reporting