VWR® E Balances
Fournisseur: VWR International
Certificats
A propos de cet article
Entry-level balances designed for basic weighing needs.
- Economical and versatile
- Large, backlit display
- Simple, five-button operation
- 5-year warranty
Sturdy construction and ease-of-use make the VWR® E-Series balance the ideal choice for the laboratory.
VWR-10001E and VWR-20001E feature an internal rechargeable battery, all other models can be powered by 6xAA batteries. RS232 interface allows connection to VWR® Dot Matrix Printer (cat. no. 10799-132).
Spécifications
- Catalog No:
- 10205-000
- 10204-996
- 10204-998
- 10204-992
- 10204-994
- 10204-990
- Operating temperature range:
- 5...40 °C
- 5...40 °C
- 5...40 °C
- 5...40 °C
- 5...40 °C
- 5...40 °C
- Plug type:
- Type-A
- Type-A
- Type-A
- Type-A
- Type-A
- Type-A
- Communication interface:
- RS232
- RS232
- RS232
- RS232
- RS232
- RS232
- Voltage:
- 100-240VAC
- 100-240VAC
- 100-240VAC
- 100-240VAC
- 100-240VAC
- 100-240VAC