VWR® C-Leg Bench Frame with Top, Double Bay Uprights and 4 Shelves
Supplier: VWR International
Certificates
About this item
VWR® lab benches are ergonomically designed to meet the needs of laboratory personnel.
- Ergonomic and highly adaptable to the needs of the user
- Flexible, modular design
- Suitable for heavy loads up to 1100 lbs.
- Offering a wide range of accessories to meet any need
There are many possible adjustments, all components fit together seamlessly, and each detail is designed for real world functionality. The lab bench adapts to the work process and the needs of the user. As a result, all movements of the user can be optimized, which in turn increases performance and productivity and decreases work-related stress. VWR® lab benches are height adjustable in manual, motorized and hand crank versions. All bench frames can be equipped with casters to create a mobile workstation solution.
Ordering information: Configurations include: frame, top, double bay uprights, 4 x 12"D Shelves. Frame color is light neutral. Top includes black phenolic or black laminate options.
Customers Who Bought This Also Bought