Human saphenous vein endothelial cells (HSaVEC), PromoCell
Fournisseur: Promocell
Certificats
A propos de cet article
Primary Human Saphenous Vein Endothelial Cells (HSaVEC) are isolated from human saphe-nous veins from single donors
- Von Willebrand factor (vWF) positive
- CD31 positive
- Smooth muscle alpha-actin negative
This product can be used in the following applications. Angiogenesis research, Atherosclerosis research, Oncology, Drug uptake studies, Cell-cell junction investigation, Inflammation studies.
The saphenous vein is often used for autotransplantation in coronary artery bypass operations, when arterial grafts are not available or many grafts are required.
Ordering information: Human Saphenous Vein Endothelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Customers Who Bought This Also Bought