Follicle Dermal Papilla Cells (HFDPC), PromoCell
Fournisseur: Promocell
Certificats
A propos de cet article
Primary Human Follicle Dermal Papilla Cells (HFDPC) are isolated from human dermis originating from the lateral scalp. Information on donor hair and skin color is available for each lot. HFDPC stain positive for alkaline phosphatase.
- Alkaline phosphatase positive
- Cryopreserved at second passage
- Available from adult lateral scalp
- Available from donors with different hair colors
Dermal papillae are embedded in a laminin and collagen IV rich extracellular matrix at the base of the hair follicles
This product can be used in the following applications. Hair growth research, Drug testing, Toxicological testing.
They are essential for the induction and maintenance of hair growth. Since they have androgen receptors, they can be used for in vitro screening of androgen blocking reagents.
Ordering information: Human Follicle Dermal Papilla Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Customers Who Bought This Also Bought