Human Umbilical Artery Smooth Muscle Cells (HUASMC), PromoCell
Fournisseur: Promocell
Certificats
A propos de cet article
Primary Human Umbilical Artery Smooth Muscle Cells (HUASMC) are isolated from the arteries of the umbilical cord and stain positive for smooth muscle α-actin
- Smooth muscle alpha-actin positive
- CD90 negative
- Routinely morphology tested
- Cryopreserved at second passage
This product can be used in the following applications. Organ specific drug testing, Cell-extracellular matrix interaction studies, Physiological and biochemical studies, Oncology, Pathological studies (e.g. hypertension).
Physiological and biochemical processes of vascular diseases can be analyzed with HUASMC in culture.
Ordering information: Human Umbilical Artery Smooth Muscle Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Customers Who Bought This Also Bought