Human Pulmonary Fibroblasts (HPF), PromoCell
Fournisseur: Promocell
Certificats
A propos de cet article
Primary Human Pulmonary Fibroblasts (HPF) are isolated from human lung tissue
- CD90 positive
- Cryopreserved at second passage
- Available from different tissues
This product can be used in the following applications. Wound healing research, Tissue engineering research, Regeneration studies, Toxicological research, Oncology, Epithelial-mesenchymal interaction studies.
They are often used for studying lung tissue repair, tissue remodeling after injury, or the response after pulmonary inflammation. Fibroblast disorders like pulmonary fibrosis or hypoxia-induced pulmonary hypertension can also be investigated with cultured HPF.
Ordering information: Human Pulmonary Fibroblasts can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Customers Who Bought This Also Bought