A propos de cet article
Primary Normal Human Dermal Fibroblasts (NHDF) are isolated form the dermis of juvenile foreskin or adult skin from different locations like the face, the breasts, the abdomen, and the thighs.
- CD90 positive
- Cryopreserved at second passage
- Available from different tissues
Normal Human Epidermal Keratinocytes (NHEK) and Normal Human Epidermal Melanocytes (NHEM) cultured in M2 medium from the same donor are available on request.) They can be used for wound healing studies and dermatological research to investigate diseases like scleroderma, fibrosarcoma, fibrosis, xeroderma pigmentosum, and histiocytoma
This product can be used in the following applications. Wound healing research, Tissue engineering research, Regeneration studies, Toxicological research, Oncology, Epithelial-mesenchymal interaction studies.
Moreover, fibroblasts are important for cancer research, tissue regeneration, and tissue engineering studies.