Human Nasal Epithelial Cells (HNEpC), PromoCell
Fournisseur: Promocell
Certificats
A propos de cet article
Primary Human Nasal Epithelial Cells (HNEpC) are isolated from normal human nasal mucosa and stain positive for cytokeratin.
- Cytokeratin positive
- Routinely morphology checked
- Cryopreserved at second passage
The epithelium in the nasal cavity cleans, humidifies, and warms inhaled air
This product can be used in the following applications. Drug uptake studies, Toxicological research, Wound healing research, Oncology, Cell differentiation studies, Respiratory disease research.
In addition, it produces mucus, which binds particles that are subsequently transported to the pharynx by cilia on the epithelial cells. HNEpC are useful for in vitro studies of processes like drug uptake studies, toxicological research, wound healing research, oncology, Cell differentiation studies, respiratory disease research.
Ordering information: Human Nasal Epithelial Cells can be ordered as cryopreserved or proliferating cells. Additional cell pellets stored in RNAlater® are available.
Customers Who Bought This Also Bought