Precision Sonic Sifter Sieves, 3", Advantec
Fournisseur: ATM
Certificats
A propos de cet article
These 3" diameter "see-through" acrylic framed sieves made with electroformed mesh are specifically designed to be used with the Sonic Sifter Separator for particles ranging from 3 µm to 150 µm
Made with electroformed precision nickel mesh meeting the current ASTM E161 sieve specifications.
Ordering information: Centerline® Sieve Inspection or Calibration Certification reports provide detailed statistical data that is linked to each serialized sieve and becomes an integral part of your Quality Assurance Program. Certifications are available as accessories.
Spécifications
- Catalog No:
- 57333-804
- 57333-826
- 57333-802
- 57333-824
- 57333-786
- 57333-808
- 57333-830
- 57333-806
- 57333-828
- 57333-790
- 57333-788
- 57333-810
- 57333-792
- 57333-814
- 57333-812
- 57333-796
- 57333-818
- 57333-794
- 57333-816
- 57333-800
- 57333-822
- 57333-798
- 57333-820
- Ø int.:
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- 3"
- Ext.Ø:
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"
- 3 1/2"