Safe-it®, Tamper Proof Labels, Bürkle
Catalog # 10217-336
Supplier: BURKLE INC
Specifications
- Description:Tamper proof labels
- Dimensions:95×20 mm
- Cat. no.:10217-336
- Supplier No.:5303-5000
Specifications
About this item
Safe-it® labels are used as protection from manipulations of samples, retention samples, for transporting samples, sealing containers with important or dangerous chemicals in the lab as well as sealing letters, shipments and more.
- With checkerboard pattern
- Visible proof of tampering
When the safe-it security seal has been broken, a silver checkerboard pattern appears on both the subsurface and surface of the sticker itself.
Customers Who Bought This Also Bought