Hand Held Scrapers, Micronova
Fournisseur: Micronova
Certificats
A propos de cet article
This hand held scraper tool is made with a seamless, one piece injection-mold that prevents bacterial growth. Designed for residue and build-up removal in clean and controlled environments.
- Injection-molded, seamless construction
- Will not scratch metal surfaces
- Made from FDA approved polypropylene
- Suitable for indirect food contact
- Handle shaped for strong grip
- Autoclavable
The scraper is chemical-resistant and will not rust or corrode.
Customers Who Bought This Also Bought