ContactUsLinkComponent
LaboPlast® Compact Spigot, Bürkle
LaboPlast® Compact Spigot, Bürkle
# de catalogue 10216-920
Fournisseur:  BURKLE INC
SPIGOT COMPACT PP 3/4IN
LaboPlast® Compact Spigot, Bürkle
# de catalogue 10216-920
Fournisseur:  BURKLE INC
Spécifications
  • Description:
    Compact spigot
  • Cat. no.:
    10216-920
  • Thread:
    3/4" inner thread cap nut
  • Supplier No.:
    0530-1000
Spécifications
A propos de cet article
The spigot Compact for canisters and barrels captivates everyone through its compact build, which makes the valve on the canister ideal for use in refrigerators and on racks, etc.
  • Space-saving design
  • Optimised discharge for even flow
  • With connecting union nut for vertical position of the stop cock
  • Tight-closing for complete seal
The short discharge does not protrude out over the lower edge of the container. PP, PE and PTFE are materials that offer outstanding chemical resistance to acids and alkalis. The Compact spigot is the ideal addition to Flachmann jerrycans for perfect storage and filling.
The discharge design creates an even stream of liquid with no splashing or spattering. The spigot features easy motion and a tight seal. It is screwed on with a connecting cap nut. This guarantees a vertical position of the valve and makes it possible to adjust the valve later on. The Compact spigot fits all containers with R 3/4" connecting threads.
Customers Who Bought This Also Bought