Sampler for Liquids, LiquiSampler, Bürkle
Fournisseur: BURKLE INC
Certificats
A propos de cet article
Fluid sampler for easy, contamination-free sampling. Perfect for sampling from open and closed barrels, vats, tanks, silos, open waters (ponds, streams, lakes, rivers).
- Disassembly and cleaning is simple
- Easy to use, one hand push-button operation
- Transparent
- Suitable for point sampling, cross-sectional sampling, bottom sampling
Made of ultra-pure and chemically inert PTFE/FEP for contamination free sampling. Fits all commercially available vats and barrels with openings of at least 25 mm diameter. Easy to clean as all surfaces are free of pores and crevices, preventing accumulation of dirt. Only round screw threads proven in foodstuffs hygiene are used. Colour coded push buttons in red, blue, green, yellow, white and black.
Customers Who Bought This Also Bought